Would you like to log in again?
Oil-X Evol Air Filters, John Henry Foster
Supplier: JOHN HENRY FOSTER MINNESOTA TE
040AA 040AO 040ACS
76246-570EA 1576.54 USD InStock
76246-570 76246-554 76246-572
Oil-X Evol Air Filters, John Henry Foster
Filters Venting Filters
Oil-X evolution genuine compressed air filter elements.
  • High efficiency coalescing
  • Dry particulate and oil vapour removal
For parker domnick hunter OIL-X EVOLUTION die-cast aluminium compressed air filters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty