Would you like to log in again?
O-Rings, Marco Rubber and Plastics
Supplier: MARCO RUBBER AND PLASTICS INC TE
345-FEP/FKM
76238-026EA 32.71 USD InStock
76238-026
O-Rings, Marco Rubber and Plastics
Seals Sealing Rings
Teflon ® encapsulated o-rings and spring energized seals offer many performance benefits in demanding environments.
  • Excellent chemical resistance
  • Temperature resistance
FEP and PFA Teflon® fluoropolymer shell delivers chemical resistance with the energizing resiliency of a rubber elastomer core combines the best of both materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty