Would you like to log in again?
Standard Outer Bag Basket, Rosedale Products
Supplier: Rosedale Products
O-730-B/PB
76240-654EA 2419.55 USD InStock
76240-654
Standard Outer Bag Basket, Rosedale Products
Baskets Safety Transporting Baskets
Standard outer bag basket, perforated, made of 304 stainless steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty