Would you like to log in again?
Overlaminate UV Protective Mats, Horizons Incorporated
Supplier: HORIZONS INCORPORATED TE
8793 8791
76242-038BX 188.7 USD InStock
76242-038 76242-040
Overlaminate UV Protective Mats, Horizons Incorporated
Seals General Purpose Sealing Films
Overlam Film does not need extra protection under normal conditions. When conditions such as abrasion, outdoor use and exposure to harsh environments occur, overlams are used to add additional protection to the processed sheets.
Uv Gloss Overlam - Recommended if the finished plate will be used in direct sunlight. It contains a UV inhibitor and it will also protect the sheet against moisture and abrasion. The finish of UV Gloss Overlam is shiny.
Matte Overlam - For applications that require a flat, non-reflective surface appearance. This overlam will also protect the sheet from moisture and abrasion..
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty