Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Petri Dish, Grainger
Supplier: Grainger
5PTF7
76244-322PK 100.81 USD InStock
76244-322
Petri Dish, Grainger
Culture Dishes Petri Dishes
Designed for optimum culture growth and media shelf life.
  • Flat design provides uniform agar thickness
  • Promotes culture growth
  • With Lid
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty