Would you like to log in again?
Vinyl Tape 471, 3M
Supplier: 3M
7000048524 7000047461
76268-066CS 3889.45 USD InStock
76268-066 76268-056
Vinyl Tape 471, 3M
Tapes
Vinyl Tape 471 is designed to resist wear, scrapes and moisture, this long-lasting tape is available in an array of vivid colors, as well as transparent.
  • Available in a wide variety of vivid colors for diverse applications
  • Good abrasion and solvent resistance for long service life
  • Rubber adhesive provides excellent holding strength on many surfaces
One-piece, clean removal from many surfaces, helps reduce clean-up and labor costs. Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty