Would you like to log in again?
ClearGreen® PVC Vinyl Tubing, Thermoplastic Biologic (TBL)
Supplier: Thermoplastic Biologic (TBL)
CG-032X062
76251-014EA 22.84 USD InStock
76251-014
ClearGreen® PVC Vinyl Tubing, Thermoplastic Biologic (TBL)
Tubing Transfer Tubing
ClearGreen® PVC vinyl tubing.
  • Non-phthalate
  • Non-Pyrogenic and Flexible
  • Non-hemolytic
ClearGreen® tubing is developed without phthalates. It has superior clarity, biocompatibility, and is friendlier to the environment at time of disposal.
Certifications: Complies with California EPA environmental standards, the EU’sREACH (article 33.1) and RoHS regulations, BSE/TSE-compliant and USP Class VI.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty