Would you like to log in again?
Pharm-A-Clear LH™ Platinum Cured Silicone Tubing, Thermoplastic Biologic
Supplier: Thermoplastic Biologic (TBL)
PLTS-C-250X437 PLTS-C-250X375 PLTS-C-250X375 PLTS-C-625X812 PLTS-C-125X250 PLTS-C-312X437 PLTS-C-500X750
76253-604EA 324.04 USD InStock
76253-604 76250-434 76250-000 76250-002 76247-232 76250-436 76253-606
Pharm-A-Clear LH™ Platinum Cured Silicone Tubing, Thermoplastic Biologic
Tubing Transfer Tubing
Pharm-A-Clear C Platinum Cured Silicone Tubing is developed to meet the most stringent requirements of the life sciences industries.
  • Medical grade
  • Superior bio compatibility
  • Extremely flexible
  • No taste or odor imparted
  • Non-pyrogenic
It is widely use for liquid media transfer and works well in peristaltic pump applications. All tubing is manufactured in a state of the art Class 100000/ISO Class 7 Clean Room environment.
Vertical extrusion techniques are employed in conjunction with continuous laser measurement to ensure the most exacting tolerances.
Certifications: Complies to U.S. Pharmacopoeia class VI, ISO 10993-5, Cytotoxicity Criteria, meets 3-A Standards and CFR Title 21 Section 177.2600.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty