Would you like to log in again?
Kapton® Polyimide Film, KR Anderson
Supplier: KR ANDERSON CO INC. TE
P221-1/2
76246-182EA 42.05 USD InStock
76246-182
Kapton® Polyimide Film, KR Anderson
Seals General Purpose Sealing Films
Suited for heavy duty, high temperature applications, ideal to use in the electrical and electronics markets.
  • Superior physical and electrical properties are retained at higher temperatures
  • Tough backing provides the ultimate in tensile strength, tear, puncture and abrasion resistance
  • High dielectric strength for critical insulating requirements
Aggressive high temperature silicone adhesive is also solvent resistant. Protection for printed circuit boards during the wave soldering or solder dipping process.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty