Would you like to log in again?
Polypropylene Core String Filters, United Filter
Supplier: UNITED FILTER TE
UP10E10P UPDN5BB10P PE5X10 UP5E30P
76239-442CS 257.32 USD InStock
76239-442 76239-440 76240-632 76239-444
Polypropylene Core String Filters, United Filter
Filters Capsule Filters
The chemical and bacterial resistance of the polyester media makes these cartridges suitable for potable water, light industrial applications, swimming pool and spa, and well water applications.
The durable non-woven polyester fabric is reusable, while being pleated to maximize its dirt holding capacity and extending the time period between changes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty