Would you like to log in again?
Air Ionizing Cartridge, Simco Ion
Supplier: Simco
92-6110A-US
76252-312EA 1867.61 USD InStock
76252-312
Air Ionizing Cartridge, Simco Ion
Ionizers
Self-contained compressed air ionizing cartridge controls static charge in production, packaging, laboratory and other environments where static build-up can cause contamination, ESD, material handling problems or microprocessor lock-up.
  • IsoStat® technology with internal air flow sensor
  • Shielded emitter points and compact size
  • Optional blow-off gun kit
  • Fits any air gun and no shock hazard
  • On-demand ionization during gun operation
  • Intrinsically balanced; no calibration needed
Adapts to compressed air lines. Compact and rugged, the cartridge can be used either for in-line ionization or as an ionizing blow-off gun.An internal sensor initiates ionization only when the gun is triggered, ensuring on-demand control of static charge.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty