Would you like to log in again?
Kelvin Probes, TESTEQUITY LLC TE
Supplier: TESTEQUITY LLC TE
6303
76251-530EA 415.09 USD InStock
76251-530
Kelvin Probes, TESTEQUITY LLC TE
Electrodes Other Electrodes
Probes for 4-wire and 2-wire measurements.
  • Patented design for high precision rapid testing
  • Gold-plated for accuracy
  • Easily move from one test point to another for rapid readings
  • For use with 4-wire Kelvin meters
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty