Would you like to log in again?
Rechargeable Battery, Newark/Element
Supplier: Newark
CUSTOM-191
76249-750EA 114.91 USD InStock
76249-750
Rechargeable Battery, Newark/Element
Batteries
This 6 Volt 1500 mAh nickel cadmium battery is used in the GS-Melcotec - KR1.0AA-4SP emergency lighting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty