Would you like to log in again?
Safety Eyewear, Loctite
Supplier: Loctite
717763
76277-070EA 159.95 USD InStock
76277-070
Safety Eyewear, Loctite
Safety Glasses
UV and visible light safety glasses protect the worker from exposure to wavelengths up to 532 nm.
  • Absorbs 99.9% of potentially harmful UVA and UVB radiation
  • Provides protection by completely absorbing visible light up to 532 nm
The operator contrast is improved and the eye finds it easier to see detail. The orange lens color filters out Visible light in the violet and blue areas of the spectrum.
Certifications: Meets ANSI specifications Z87.1 / CSA Z 94.3.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty