Would you like to log in again?
Journey® Safety Eyewear, Radians
Supplier: Radians
JR0110ID
76238-860EA 10.6 USD InStock
76238-860
Journey® Safety Eyewear, Radians
Safety Glasses
Journey® safety glasses provide excellent optics and coverage. Scratch resistant coating on all protective lenses.
  • 4-position telescoping temples for superior fit
  • 9.75 base lens provides excellent optics and coverage
  • Rubber nose piece allows for all day comfort
All lenses are hard coated for scratch resistance. Ballistic Testing. Provides 99.9% UVA/UVB protection. Eyewear also available with Realtree Hardwoods® camo pattern on the frame.
Certifications: Safety glasses meet ANSI Z87.1 + standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty