Would you like to log in again?
Safetyflex® Apron, Conney Safety Products
Supplier: Conney Safety
159782
76243-486EA 119.32 USD InStock
76243-486
Safetyflex® Apron, Conney Safety Products
Workwear
The SafetyFlex® flame resistant specialty PVC on polyester apron combines tough protection and comfortable fit.
  • Made to resist industrial acids, chemicals, and caustics
  • Flame resistant tested
  • Self extinguishing with flameout
The heavy exterior coating offers excellent permeation performance and liquid splash protection. Equipped with one strap around the waist and one around the neck with a buckle for a secure and adjustable fit.
Afterglow less than or equal to 2 seconds after removal of ignition source and char length less than 6 inches.
Certifications: Complies to ASTM D6413.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty