Would you like to log in again?
Wetordry™ Abrasive Sheet, 3M
Supplier: 3M
7000118294
76244-372CS 1032.31 USD InStock
76244-372
Wetordry™ Abrasive Sheet, 3M
Abrasives
Wetordry™ Abrasive Sheet 413Q is an abrasive sanding paper constructed on a light, weight paper backing and features a waterproof resin bonding to help prevent heat buildup.
  • Silicon carbide mineral provides an aggressive cut and a consistent, fine finish
  • Waterproof material allows for easy debris removal
  • Can be easily cut and folded to fit any hand tool or work piece crevice
The silicon carbide abrasive and waterproof backing make this paper an ideal product for sanding applications such as paint prep, sanding, sealer sanding, and solid surface finishing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty