Would you like to log in again?
Scotch® Tape, Printable, 3M
Supplier: 3M
Scotch®
7000047439 7010292867 7000047440
76242-349CS 2900.13 USD InStock
76242-349 76245-622 76242-388
Scotch® Tape, Printable, 3M
Tapes
Scotch® printable flatback paper tape features a rubber adhesive that instantly adheres to a wide variety of surfaces.
  • Printable with standard flexographic inks and easily markable
  • Adhesive bonds to wide variety of paper, film, fabric and yarn surfaces
  • Creates good paint lines when used for masking purposes
  • Conforms to corners, curves, and irregular surfaces
It detaches easily and provides clean removal from a variety of substrates allowing it to be used in painting applications. The flatback paper backing is printable and features good tensile strength. This tape is ideal for use in temperatures up to 93 °C (200 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty