Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum-Cured Silicone Tubing STHT-C, Holland Applied Technologies

Supplier: Holland Applied Technologies
STHT-C-030-0F-BULK
76252-106CS 240.87 USD InStock
76252-106
Platinum-Cured Silicone Tubing STHT-C, Holland Applied Technologies
Tubing Transfer Tubing
Sani-Tech STHT-C is an ultra-pure biopharmaceutical-grade tubing that has a use life unmatched by any other known process tubing.

  • STHT-C Tubing is ultra-pure biopharmaceutical-grade silicone
  • Unsurpassed surface smoothness
  • STHT-C is autoclavable and sterilizable
  • Temperature range from –80 °F (–62 °C) to 500 °F (260 °C)
  • Laser-marked part numbers and lot codes (¹/₁₆'' int.Ø andgreater)
  • Color striping available

Sani-Tech STHT-C platinum-cured silicone tubing resists temperature extremes, ozone, radiation, oisture, compression sets, weathering and chemical attack, and imparts no odors to fluids transported within it.

These tubing withstands repeated autoclaving and sterilization methods and resists the adherence of blood products and other sanitary fluids.

Certifications: Sani-Tech STHT-C Meets USP Class VI, European Pharmacopoeia 3.1.9, and FDA criteria.

Ordering information: Available in multiple length coils.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty