Would you like to log in again?
Edsyn Lifter ESD-Safe Portable Vacuum Tool, Jensen Tools
Supplier: Jenson Tools
LP200
76248-808EA 126.05 USD InStock
76248-808
Edsyn Lifter ESD-Safe Portable Vacuum Tool, Jensen Tools
Soldering Supplies Desoldering Tools
The 3-ounce pen sized tool is ideal for picking, placing and removing flat components up to 500 grams and accepts wide variety of needles and suction cups. Comes complete, small angled lifting needle with 1/8" static-safe silicone suction cup, angled lifting needle with 1/4" static-safe silicone suction cup, large angled lifting needle with 3/8" static-safe silicone suction cup.
  • ESD-safe
  • Lightweight
  • Portable type
Lifting capacity: 500 grams
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty