Would you like to log in again?
Disposable Specimen Containers, Dynalon
Supplier: Dynalon
453595 453605
76244-028CS 540.8 USD InStock
76244-028 76244-030
Disposable Specimen Containers, Dynalon
Sample Containers General Purpose Containers
These containers are inert and cannot be attacked by formaldehyde or weak acids or bases.
  • Resist freezing or boiling
  • Have long shelf lives and are easily disposable
  • Sturdy enough for reuse
  • Easy to store
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty