Would you like to log in again?
ESD-Safe Spray Mister Bottle, Testequity LLC TE
Supplier: TESTEQUITY LLC TE
SMB-2-ESD
76239-824CS 1477.45 USD InStock
76239-824
ESD-Safe Spray Mister Bottle, Testequity LLC TE
Bottles Spray Bottles
Spary bottle to deliver a fine mist with this trigger-type pump and ESD-safe bottle.
These ESD-safe bottles have an average surface resistivity of 10⁹ to 10¹⁰ and can dissipate a charge of ±5000 V in less than 2 s.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty