Would you like to log in again?
Spring Balances, ASG
Supplier: ASG, Division of Jergens
64022 64021
76245-132EA 89.65 USD InStock
76245-132 76245-134
Spring Balances, ASG
Balances and Scales
Spring tool balancers with either 1.0 to 3.0 lb. or 3.0 to 6.0 lb. capacity.
  • Reduce operator fatigue
  • Tools become weightless
  • Smooth operation
  • Tool is raised to the up position when released
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty