Would you like to log in again?
Disposal Cabinet, PALBAM
Supplier: Palbam
GDC-8040
76277-164EA 6829.48 USD InStock
76277-164
Disposal Cabinet, PALBAM
Bins Waste Bins
This disposal cabinet can be used on its own as a disposal/recycle unit or it can be combined with dispensers to create a dispensing/disposal unit.
  • Electropolished type 304 stainless steel construction
  • Two waste throw openings, two doors and leveling feet
  • Includes two removable waste bins (plastic) for easy emptying
Can be used in cleanroom environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty