Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Teledyne TW-1 Thermal Stripper, TESTEQUITY LLC TE
Supplier: TESTEQUITY LLC TE
TW-1
76242-290EA 2463.05 USD InStock
76242-290
Teledyne TW-1 Thermal Stripper, TESTEQUITY LLC TE
Cutters
This versatile model is designed to remove a variety of insulation types, up to 1.5 inches, from single-strand, multi-strand, or shielded wire.
The TWC-1 works well on Teflon, Kapton, Nylon, Vinyl and Neoprene.
Delivery information: Supplied with notched blades, for finer wire stripping to 36AWG use the blank blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty