Would you like to log in again?
DermAssist® Surgical Gloves, Innovative Healthcare
Supplier: Innovative Healthcare Corporation
133800 133900 133650
76237-862CS 298.04 USD InStock
76237-862 76237-860 76237-858
DermAssist® Surgical Gloves, Innovative Healthcare
Gloves
These low-protein, powder-free gloves eliminate air-borne particles of latex protein associated with powdered surgical gloves.
  • Improved wet or dry grip
  • Soft, more comfortable fit
  • Reduced chances of latex allergic reactions
Ultra-low modulus is designed for greater comfort, less hand fatigue during long surgical procedures.
Certifications: ASTM D3577, ASTM D5151, ASTM F1671 and ASTM D5712 tested.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty