Would you like to log in again?
Swing Sleeves, Ansell
Supplier: Ansell
950504
76238-262CS 252.82 USD InStock
76238-262
Swing Sleeves, Ansell
Disposable Garments
This polyethylene sleeve is designed to protect part of the body against splashes from certain chemicals.
  • Excellent low-cost protection
  • Protects against punctures, snags and abrasions
  • Provides resistance to chemicals, fats, oils and grease
  • Elasticized sleeves opening are completely sealed with no exposed elastic
  • Heat sealed, with elastics at the extremities
Caution: The elastic band on this product may contain natural rubber latex which may cause allergic reactions in some individuals.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty