Would you like to log in again?
Syringe Barrels and Piston Sets, EFD
Supplier: Nordson EFD
7012153 7012096 7012114 7012091 7012074 7360577 7012075 7012085 7012136 7012118 7012149
76247-646PK 118.26 USD InStock
76247-646 76241-722 76267-510 76245-646 76241-166 76238-214 76241-010 76247-468 76245-488 76252-438 76243-808
Syringe Barrels and Piston Sets, EFD
Syringe Barrels, Plungers and Pistons
Precision fit between syringe and piston ensures consistent fluid deposits.
  • Silicone-free
  • Package labels include lot numbers for process control and traceability
Wiper piston improves fluid control, keeps fluids from dripping and eliminates waste by wiping the syringe wall clean.
Sets are available in clear for most fluids, transparent amber for UV and light-sensitive materials (up to 550 nm) and opaque black for complete light blockage.
Delivery information: Each box contains one resealable bag of syringes and SmoothFlow™ pistons.
Packaging: Dust-free packaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty