Would you like to log in again?
IPG™ BD1 Paper Tapes, Hisco
Supplier: Hisco
BD1-WHT-6MMX2000-3600
76246-812EA 270.64 USD InStock
76246-812
IPG™ BD1 Paper Tapes, Hisco
Tapes
A high strength, rope fiber flatback paper, pressure sensitive tape featuring high tensile strength with good edge tear resistance.
  • Excellent mass to mass adhesion
Ideal for bandoliering (axial taping) electronic components, sequential taping of electronic components, tape reeling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty