Would you like to log in again?
Torque Arms, ASG
Supplier: ASG, Division of Jergens
65050
76251-818EA 3615.61 USD InStock
76251-818
Torque Arms, ASG
Support Stands
Easily mounts or clamps to bench-top (clamp optional).
  • Maintains tool location while allowing easy movement
  • Durable, lightweight
  • Adjustable height
  • Maximum arm reach 27.5"
Helps reduce potential operator injury to arm, wrist, or hand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty