Would you like to log in again?
Tryptose Soy Broth, USP, Neogen
Supplier: Neogen
NCM0004B 7164-B
76252-142EA 1358.39 USD InStock
76252-142 76238-302
Tryptose Soy Broth, USP, Neogen
Media for Microbiology Prepared Media for Microbiology
Tryptic soy broth is used for the cultivation of a wide variety of microorganisms in a laboratory setting.
  • Final pH: 7.3±0.2 at 25 °C
With the addition of 6.5% NaCl, TSB can be used for the selective growth of group D streptococci.
Recommended for testing bacterial contaminants in cosmetics and complies with established standards in the food industry.
Caution: Not intended for use in the diagnosis of disease or other conditions in humans.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty