Would you like to log in again?
Carbofib 3-CF Tweezers, Aven Tools
Supplier: Aven
18702
76243-982EA 117.74 USD InStock
76243-982
Carbofib 3-CF Tweezers, Aven Tools
Forceps
These Swiss made, ESD safe tweezers feature excellent wear and abrasion resistance.
  • High rigidity, excellent tensile and flexural strength
  • Good chemical resistance to oils, grease, fuels, and nonpolar solvents
Ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass and ceramic substrates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty