Would you like to log in again?
Omega Supreme Electronic Vacuum Cleaner, VACOSE110, Jensen Tools
Supplier: Jenson Tools
VACOSE110
76252-912EA 1248.72 USD InStock
76252-912
Omega Supreme Electronic Vacuum Cleaner, VACOSE110, Jensen Tools
Vacuum Cleaners
This vacuum cleaner features 10' ESD safe stretch hose and a set of hose end plugs to keep the inside of the vacuum clean during transport and storage.
  • Less than 70 decibel
  • Quiet and powerful
  • Self storage compartment
  • ESD-Safe
  • Uses 20% less energy
Plug each end of the hose before storing in the lid. Toner, dust and dirt does not spill out from the stretch hose.
Delivery information: Includes vacuum with quiet 1 HP motor, 10' ESD-safe stretch hose, ESD-safe 16" rubber flex neck tool, ESD crevisce tool, electronic line filter, standard cartridge filter, ESD-safe brush, ESD safe needle nozzle, 10' detachable power cord, shoulder strap with d-rings.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty