Would you like to log in again?
3M™ Tapes Vinyl , 3M
Supplier: 3M
7000047467 7000028852 7000047469
76268-368CS 683.94 USD InStock
76268-368 76243-946 76248-242
3M™ Tapes Vinyl , 3M
Tapes
These tapes stick on contact to a wide variety of surfaces for fast application with no dripping, drying, or cleanup necessary, and removes cleanly, leaving no sticky residue.
  • Maintains vivid colour even when exposed to abrasion and solvents
  • Rubber adhesive provides excellent holding strength on many surfaces
  • Allows fast and easy application to a variety of surfaces
  • Easy removal with no adhesive transfer makes cleanups efficient
Commonly used for marking hazards and aisles, colour-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.
Vinyl tape is highly conformable, making it easy to apply to curved areas as well as rough or uneven surfaces and can be used in temperatures ranging from 40 to 170 °F. Vinyl tapes are used to clearly mark lanes, corridors and hazardous or no-go areas in factories, warehouses, and hospitals. Using vinyl tape for lane marking instead of painting eliminates the need to ventilate paint solvents and since vinyl tapes can be removed quickly and cleanly, they are also a faster, more versatile, and less costly option than painting. These rugged tapes are also used in automotive maintenance and repair, construction, marine, commercial vehicle and other industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty