Would you like to log in again?
QbE® Cleaning Platform, ITW Chemtronics
Supplier: ITW Chemtronics
QBE
76244-330BX 20.86 USD InStock
76244-330
QbE® Cleaning Platform, ITW Chemtronics
Wipes Cleaning Kits and Pads
QbE® cleaning platform offers a more complete removal of microscopic contaminants, without the chance of damage to the optical fiber end face.
  • Complete fiber optic cleaning system
  • Effective wet or dry connector cleaning
  • Provides ideal cleaning surface
  • Portable cleaning system easily fits on work benches
Heavy duty lint-free wiping material is tough enough to remove buffer-gel and soft enough for all end-face cleaning.
Self-contained cleaning tools that can be used dry or wet, they are convenient and economical for use in field or OEM applications. Other applications include end-face connector cleaning for SC, ST and FC connectors, splice preparation and uffer gel removal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty