Would you like to log in again?
Sta-Strap® Cable Ties, Panduit
Supplier: Panduit
SST1.5M-M SSM2S-C
76242-666PK 168.28 USD InStock
76242-666 76250-868
Sta-Strap® Cable Ties, Panduit
Ties
Marker tie features two-piece design, used to fasten and identify bundles at the same time and can be custom imprinted.
Cable tie features two-piece design, extremely low thread force, may be released prior to final tensioning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty