Would you like to log in again?
Scotch-Brite™ Clean and Finish Discs, RS Hughes
Supplier: RS HUGHES TE
Scotch-Brite™
048011-91932
76250-532CS 562.41 USD InStock
76250-532
Scotch-Brite™ Clean and Finish Discs, RS Hughes
Abrasives
Scotch-Brite™ clean and finish discs combine conformable material and abrasive mineral to effectively blend, clean, and polish surfaces on a wide variety materials such as aluminum, stainless steel, plastics, wood, and fiberglass; producing a clean, consistent finish without undercutting or damaging the base material.
  • Soft abrasive mineral and non-woven web are ideal for cleaning, blending, finishing, and light coating and corrosion removal
  • Non-woven 3D structure provides long life and consistent finishes
  • Attaches to random orbital sander (hookit backup pad) for increased cut-rate and cover larger areas
  • Non-woven web is load and heat resistant
Conformable pad gets into corners and over contoured surfaces to produce a uniform finish. Open web construction resists loading and rinses clean for repeated use.
They feature an abrasive mineral that blends, finishes, and polishes metals, composites and advanced surfaces and is an effective abrasive tool for paint preparation, scuffing, and light corrosion removal.
Ideal for finishing, paint preparation, surface blending, and scuffing application.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty