Would you like to log in again?
Aquatrex Large Diameter Cartridge Filter, PFC Equipment
Supplier: PFC EQUIPMENT INC. TE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
LD20-20
76238-876CS 402.14 USD InStock
76238-876
Aquatrex Large Diameter Cartridge Filter, PFC Equipment
Filters Filter Cartridges
Aquatrex filter combines innovative depth filter technology, quality cost effectiveness to provide excellent filtration value in residential and light commercial applications.
  • Unique dual-graded density design of the extended media captures particles throughout entire filter depth
  • High dirt-holding capacity means longer life and fewer change-outs
  • Precision final filter results in consistent highperformance filtration
  • Large media area allows for high flow rates
Applications include whole house filtration, spa and pool filtration, restaurant and food service and water filtration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty