Would you like to log in again?
Xcelite® Allen® Hex Socket Type Ballpoint Style Screwdrivers, Apex
Supplier: Apex
Xcelite®
XC-LN764BPN LN23BPN
76242-428EA 33.28 USD InStock
76242-428 76242-430
Xcelite® Allen® Hex Socket Type Ballpoint Style Screwdrivers, Apex
Screwdrivers
These screwdrivers feature unique “ballpoint” tip which makes Xcelite® drivers ideal for tightening and removing Allen type socket head screws in out-of-reach places or merely when wanting an easier, quicker way to drive them home.
  • Will engage screw head from any angle
Slips easily into sockets and holds securely without reaming, because ends are precision-machined to close tolerance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty