Would you like to log in again?
Fanfold Label, Z-Select, Zebra
Supplier: Zebra
Z-Select
10020064
76249-788EA 171.03 USD InStock
76249-788
Fanfold Label, Z-Select, Zebra
Labels
This Zebra label is a premium direct thermal paper label with an all-temp permanent acrylic adhesive.
  • Recommended for indoor use
  • survives temperatures as low as –65 °F and can be applied at temperatures as low as –20 °F
This Zebra label is specially coated to provide optimal performance in a variety of environments.
Applications include labeling of packaging material including corrugate, plastic, and metal, product identification, compliance labeling work in process labeling, food labeling.
Certifications: Meets FDA 175.105 indirect food contact requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty