Would you like to log in again?
Barnes Dropping Bottle, United Scientific Supplies
Supplier: United Scientific Supplies
BOT030-36-VWR
76293-356PK 76.62 USD InStock
76293-356
Barnes Dropping Bottle, United Scientific Supplies
Bottles Dropping and Dispensing Bottles
The wide, square body provides extra stability and efficient storage.
  • Sturdy glass construction
  • Rubber stopper with glass pipet closure included
Square-shaped, clear flint glass bottles with glass pipet and rubber bulb stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty