Would you like to log in again?
4-Prong Extension Clamp with Boss Head, United Scientific Supplies
Supplier: United Scientific Supplies
CLFCORK2-VWR
76293-390EA 26.5 USD InStock
76293-390
4-Prong Extension Clamp with Boss Head, United Scientific Supplies
Clamps General Purpose Clamps
The 4-prong die-cast clamp is attached to a chrome-plated steel rod.
  • Corked lined Jaws
  • Die cast clamp
Jaws are lined with cork. Overall length is 8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty