Would you like to log in again?
C-Clamps, United Scientific Supplies
Supplier: United Scientific Supplies
CLMPC3-VWR CLMPC5-VWR CLMPC2-VWR CLMPC4-VWR CLMPC6-VWR
76293-420EA 4.89 USD InStock
76293-420 76293-422 76293-516 76293-514 76293-518
C-Clamps, United Scientific Supplies
Clamps General Purpose Clamps
Securely hold two flat object together.
  • Cast iron frame
  • Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty