Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNAP25 Rabbit Polyclonal Antibody

Supplier: Sino Biological
11585-RP01-100 11585-RP01-400 11585-RP01-200
103634-486EA 270.2 USD InStock
103634-486 103634-488 103634-490
Anti-SNAP25 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human SNAP25 (rh SNAP25; Catalog#11585-H07E; P60880-1; Met 1-Gly 206). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SNAP-25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty