Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MUC1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
12123-T24-50 12123-T24-100 12123-T24-200
103587-604EA 142.96 USD InStock
103587-604 103609-160 103609-162
Anti-MUC1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human MUC-1 / Mucin 1 / CD227 (rh MUC-1 / Mucin 1 / CD227;NP_001018016.1; Met1-Gly167). MUC-1 / Mucin 1 / CD227 specific IgG was purified by Human MUC-1 / Mucin 1 / CD227 affinity chromatography.

Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty