Would you like to log in again?
Burette Clamp with Boss Head, Coated Jaws, United Scientific Supplies
Supplier: United Scientific Supplies
COBR3-FR-VWR
76293-406EA 8.96 USD InStock
76293-406
Burette Clamp with Boss Head, Coated Jaws, United Scientific Supplies
Clamps General Purpose Clamps
Excellent for holding small objects.
  • PVC coated Jaws
  • Jaws can be rotated
The jaws open from 15mm to 45 mm (0.5" to 1.75"). The clamp attaches to rods up to 13mm (0.5") O.D. Jaws can be rotated and locked at any angle. Jaws are covered with PVC sleeves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty