Would you like to log in again?
3-Prong Universal Clamp, with Holder, United Scientific Supplies
Supplier: United Scientific Supplies
ECBH01-VWR
76293-374EA 32.31 USD InStock
76293-374
3-Prong Universal Clamp, with Holder, United Scientific Supplies
Clamps General Purpose Clamps
Useful clamp that can be adjusted to any need.
  • Adjustable to any angle
  • Connects to any standard support rod
Clamp features vinyl coated 3-prong jaws that open up to 2.25" wide. Includes fiberglass sleeves and a clamp holder that can be rotated 360°.
Delivery information: Includes fiberglass sleeves and a clamp holder that can be rotated 360°.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty