Would you like to log in again?
Nickel Crucibles, United Scientific Supplies
Supplier: United Scientific Supplies
NCR025-VWR
76293-528EA 23.06 USD InStock
76293-528
Nickel Crucibles, United Scientific Supplies
Crucibles
Unibody construction from sheet nickel, polished inside and out.
  • Polished inside and out
  • Includes a cover
These nickel crucibles can withstand temperatures up to 600 °C.
Delivery information: Covers included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty