Would you like to log in again?
Slide Holding Forceps, United Scientific Supplies
Supplier: United Scientific Supplies
SFK001-VWR
76293-304EA 1.64 USD InStock
76293-304
Slide Holding Forceps, United Scientific Supplies
Forceps
Safety hold glass slides.
  • Holds all sizes of slides gently
  • Plated steel wire
Hold your slide without having your hands touch the slide. Made of plated steel wire for careful and tight handling of cover glasses and slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty