Would you like to log in again?
Turboshield™ Face Protection Shield, Uvex
Supplier: Uvex
Turboshield™
S9550
10027-348EA 36.34 USD InStock
10027-348
Turboshield™ Face Protection Shield, Uvex
Masks Hygiene Masks
Turboshield™ PC Visor face protection system provides superior comfort for longer wear.
  • Easiest visor exchange available
  • Optimal balance when used with a hard hat
Ideal for workers exposed to falling or flying objects, impact, splash, and airborne debris; it can also be comfortably worn with most goggles, respirators and earmuffs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty